missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane 98.0+%, TCI America™

Catalog No. D0134500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0134500G 500 g
D013425G 25 g
D0134100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0134500G Supplier TCI America Supplier No. D0134500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

Melting Point 155°C
Color White-Yellow
Boiling Point 174°C
Quantity 500 g
UN Number 3263
Formula Weight 112.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane
EINECSNumber (5)-1141
RTECSNumber HM0354200
TSCA Yes