missing translation for 'onlineSavingsMsg'
Learn More

1,4-Diazabicyclo[2.2.2]octane, 98%

Catalog No. AAA1400322
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1400322 100 g
AAA1400314 25 g
AAA1400336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1400322 Supplier Thermo Scientific Chemicals Supplier No. A1400322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 280-57-9 | C6H12N2 | 112.176 g/mol

1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.

Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 280-57-9
Molecular Formula C6H12N2
Molecular Weight (g/mol) 112.176
MDL Number MFCD00006689
InChI Key IMNIMPAHZVJRPE-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane, triethylenediamine, dabco, dabco 33lv, 1,4-ethylenepiperazine, dabco crystal, teda, texacat td 100, dabco s-25
PubChem CID 9237
IUPAC Name 1,4-diazabicyclo[2.2.2]octane
SMILES C1CN2CCN1CC2

Specifications

Melting Point 155°C to 160°C
Density 1.14
Boiling Point 174°C to 176°C
Flash Point 62°C (143°F)
Odor Amine-like
Quantity 100 g
UN Number UN1325
Beilstein 103618
Sensitivity Hygroscopic
Merck Index 14,9669
Solubility Information Soluble in water,acetone,benzene,ethanol and methyl ethyl ketone.
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 1,4-Diazabicyclo[2.2.2]octane
Show More Show Less
Hazard Category H228-H302-H315-H318-H500
Hazard Statement GHS H Statement
H228-H318-H302-H335-H315
Flammable solid.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 205-999-9
RTECSNumber HM0354200
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only