missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedimethanol, cis + trans, 99%

Catalog No. AAA1676936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1676936 500 g
AAA1676922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1676936 Supplier Thermo Scientific Chemicals Supplier No. A1676936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105-08-8 | C8H16O2 | 144.21 g/mol

CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

Solubility
Miscible with water, glycolether and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 105-08-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00001512,MFCD00066360
InChI Key YIMQCDZDWXUDCA-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis
PubChem CID 7735
IUPAC Name [4-(hydroxymethyl)cyclohexyl]methanol
SMILES OCC1CCC(CO)CC1

Specifications

Melting Point ∼30°C
Density 1.04
Boiling Point 282°C to 284°C
Flash Point 161°C (322°F)
Quantity 500 g
Beilstein 1902271
Solubility Information Miscible with water,glycolether and ketones.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 1,4-Cyclohexanedimethanol, cis + trans
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 203-268-9
RTECSNumber GU9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only