missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedicarboxylic acid, cis + trans, 98%

Catalog No. AAB2312536
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2312536 500 g
AAB2312522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2312536 Supplier Thermo Scientific Chemicals Supplier No. B2312536
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1076-97-7 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1076-97-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00001465
InChI Key PXGZQGDTEZPERC-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane
PubChem CID 14106
IUPAC Name cyclohexane-1,4-dicarboxylic acid
SMILES C1CC(CCC1C(=O)O)C(=O)O

Specifications

Melting Point >300°C
Flash Point 235°C (455°F)
Quantity 500 g
Beilstein 1870377
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material 1,4-Cyclohexanedicarboxylic acid, cis + trans
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 214-068-6
RTECSNumber GU9060000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only