missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanediamine (cis- and trans- mixture) 97.0+%, TCI America™
Spécifications
Spécifications
| Color | White-Yellow |
| Boiling Point | 197°C |
| Quantity | 5 mL |
| UN Number | 3259 |
| Formula Weight | 114.19 |
| Percent Purity | ≥97.0% (GC,T) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 1,4-Cyclohexanediamine (cis- and trans- mixture) |
Sécurité et manipulation
| RTECSNumber | GU8750500 |
| TSCA | No |