missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanebis(methylamine), cis + trans, 96%

Catalog No. AAL1340714
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1340714 25 g
AAL1340703 1 g
AAL1340706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1340714 Supplier Thermo Scientific Chemicals Supplier No. L1340714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2549-93-1 | C8H18N2 | 142.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2549-93-1
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00039463
InChI Key OXIKYYJDTWKERT-UHFFFAOYSA-N
Synonym 1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanedimethanamine, bamch, cis-1,4-bis aminomethyl cyclohexane, 1,4-cyclohexanebis methylamine, cyclohexane-1,4-diyldimethanamine, trans-1,4-bis aminomethyl cyclohexane, 4-aminomethyl cyclohexyl methanamine, trans-1,4-cyclohexanedimethanamine, cyclohex-1,4-ylenebis methylamine
PubChem CID 17354
IUPAC Name [4-(aminomethyl)cyclohexyl]methanamine
SMILES C1CC(CCC1CN)CN

Specifications

Density 0.95
Boiling Point 239°C to 244°C
Flash Point 106°C (222°F)
Refractive Index 1.49
Quantity 25 g
UN Number UN2922
Beilstein 2071383
Sensitivity Air Sensitive
Formula Weight 142.25
Percent Purity 96%
Chemical Name or Material 1,4-Cyclohexanebis(methylamine), Cis + Trans
Show More Show Less
Hazard Category H302+H312-H314
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 219-840-6
RTECSNumber GU7000010
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only