missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanesultone, 99%

Catalog No. AAL0399222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0399222 100 g
AAL0399214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0399222 Supplier Thermo Scientific Chemicals Supplier No. L0399222

CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
1,4-Butanesultone is an alkylating agent used in synthetic chemistry. It serves as a wood preservative. It is employed as an intermediate of the organic sulfonation agent. Further, it is used as a raw material for electroplating intermediate. In addition to this, it finds application in active pharmaceutical ingredients, photographic materials, lithium battery and surface active agents.

Solubility
Miscible with chloroform and methanol. Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 1633-83-6
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.17
MDL Number MFCD00006584
InChI Key MHYFEEDKONKGEB-UHFFFAOYSA-N
Synonym 1,4-butane sultone, 1,4-butanesultone, butane sultone, butanesultone, 1,2-oxathiane, 2,2-dioxide, butanesulfone, 1,2-oxathiane 2,2-dioxide, 1,4-butylene sulfone, delta-valerosultone, 1,4-butanesulfone
PubChem CID 15411
IUPAC Name oxathiane 2,2-dioxide
SMILES O=S1(=O)CCCCO1

Specifications

Melting Point 12°C to 15°C
Density 1.335
Boiling Point 114°C to 116°C (1.5 mmHg)
Flash Point 153°C (307°F)
Refractive Index 1.464
Quantity 100 g
Beilstein 110588
Sensitivity Moisture sensitive
Solubility Information Miscible with chloroform and methanol. Slightly miscible with water.
Formula Weight 136.17
Percent Purity 99%
Chemical Name or Material 1,4-Butanesultone
Show More Show Less
Hazard Category H302-H315-H320-H335-H341-H351
Hazard Statement GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 216-647-9
RTECSNumber RP4300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only