missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trifluoromethyl)benzene, 99%

Catalog No. p-7024158
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1841122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1841122 Supplier Thermo Scientific Chemicals Supplier No. A1841122

CAS: 433-19-2 | C8H4F6 | 214.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 433-19-2
Molecular Formula C8H4F6
Molecular Weight (g/mol) 214.11
MDL Number MFCD00000402
InChI Key PDCBZHHORLHNCZ-UHFFFAOYSA-N
Synonym 1,4-bis trifluoromethyl benzene, 1,4-di trifluoromethyl benzene, 1,4-bis trifluoromethyl-benzene, hexafluoro-p-xylene, p-bis trifluoromethyl benzene, benzene, 1,4-bis trifluoromethyl, 1,4-bis-trifluoromethylbenzene, p-trifluoromethylbenzotrifluoride, ptf-tfm, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-p-xylene
PubChem CID 67945
IUPAC Name 1,4-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point -1°C
Density 1.381
Boiling Point 116°C to 117°C
Flash Point 21°C (69°F)
Odor Aromatic
Refractive Index 1.379
Quantity 100 g
UN Number UN1993
Beilstein 1912445
Formula Weight 214.11
Percent Purity 99%
Chemical Name or Material 1,4-Bis(trifluoromethyl)benzene
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 207-086-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only