missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(phenylethynyl)benzene, 97%

Catalog No. AAH3039514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3039514 25 g
AAH3039503 1 g
AAH3039506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3039514 Supplier Thermo Scientific Chemicals Supplier No. H3039514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1849-27-0 | C22H14 | 278.354 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1849-27-0
Molecular Formula C22H14
Molecular Weight (g/mol) 278.354
MDL Number MFCD00160829
InChI Key FPVSTPLZJLYNMB-UHFFFAOYSA-N
Synonym 1,4-bis phenylethynyl benzene, 1,4-bis 2-phenylethynyl benzene, benzene,1,4-bis 2-phenylethynyl, p-bis phenylethynyl benzene, 1,4-bis phenylethynyl benzene #, 1,4-bis 2-phenyl-ethynyl-benzene
PubChem CID 624226
IUPAC Name 1,4-bis(2-phenylethynyl)benzene
SMILES C1=CC=C(C=C1)C#CC2=CC=C(C=C2)C#CC3=CC=CC=C3

Specifications

Melting Point 176°C to 178°C
Quantity 25 g
Formula Weight 278.36
Percent Purity 97%
Chemical Name or Material 1,4-Bis(phenylethynyl)benzene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only