missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(3-aminopropyl)piperazine, 98%

Catalog No. AAL1250822
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1250822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1250822 Supplier Thermo Scientific Chemicals Supplier No. L1250822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7209-38-3 | C10H24N4 | 200.33 g/mol

1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(3-aminopropyl)piperazine was used to functionalize natural montmorillonite (M) and synthetic kanemite (K)1.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7209-38-3
Molecular Formula C10H24N4
Molecular Weight (g/mol) 200.33
MDL Number MFCD00006161
InChI Key XUSNPFGLKGCWGN-UHFFFAOYSA-N
Synonym 1,4-bis 3-aminopropyl piperazine, 1,4-piperazinedipropanamine, 3,3'-piperazine-1,4-diyl bis propan-1-amine, bis aminopropyl piperazine, n,n'-bis 3-aminopropyl piperazine, unii-7gvx6c734b, bapp, ccris 8915, 1,4-di 3-aminopropyl piperazine, 3,3'-piperazine-1,4-diyldipropan-1-amine
PubChem CID 81629
IUPAC Name 3-[4-(3-aminopropyl)piperazin-1-yl]propan-1-amine
SMILES C1CN(CCN1CCCN)CCCN

Specifications

Melting Point 14°C
Density 0.973
Boiling Point 150°C to 152°C (2 mmHg)
Flash Point 162°C (323°F)
Refractive Index 1.5018
Quantity 100 g
UN Number UN2735
Beilstein 111747
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 200.33
Percent Purity 98%
Chemical Name or Material 1,4-Bis(3-aminopropyl)piperazine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 230-589-1
RTECSNumber TK8700000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only