missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Bis[(1H-imidazol-1-yl)methyl]benzene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 132°C |
| Color | White |
| Quantity | 1 g |
| Synonym | alpha,alpha′C-Bis[(1-imidazolyl)methyl]-p-xylene |
| Formula Weight | 238.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,4-Bis[(1H-imidazol-1-yl)methyl]benzene |
Safety and Handling
| TSCA | No |