missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzoquinone Dioxime 95.0+%, TCI America™

Catalog No. B009025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B009025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B009025G Supplier TCI America Supplier No. B009025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 105-11-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00063636
InChI Key LNHURPJLTHSVMU-CGXWXWIYSA-N
Synonym 1,4-benzoquinone dioxime, p-benzoquinonedioxime, p-benzoquinone dioxime, p-quinone dioxime, quinone dioxime, benzoquinone dioxime, p-quinone oxime, actor q, dioxime p-benzoquinone, dibenzo pqd
PubChem CID 7737
IUPAC Name N-[(1E,4E)-4-(hydroxyimino)cyclohexa-2,5-dien-1-ylidene]hydroxylamine
SMILES ON=C1C=CC(C=C1)=NO

Specifications

Quantity 25 g
Formula Weight 138.13
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzoquinone Dioxime
EINECSNumber (3)-1009
RTECSNumber DK4900000
TSCA Yes