missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane 98.0+%, TCI America™

Catalog No. B119825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B119825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B119825G Supplier TCI America Supplier No. B119825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 493-09-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00006821
InChI Key BNBQRQQYDMDJAH-UHFFFAOYSA-N
Synonym 1,4-benzodioxane, 1,4-benzodioxan, 2,3-dihydrobenzo b 1,4 dioxine, 1,4-benzodioxin, 2,3-dihydro, pyrocatechol ethylene ether, 1,2-ethylenedioxy benzene, benzodioxan, ethylene o-phenylene dioxide, 1,2-ethylenedioxybenzene, 1, 4-benzdioxan
PubChem CID 10301
IUPAC Name 2,3-dihydro-1,4-benzodioxine
SMILES C1COC2=CC=CC=C2O1

Specifications

Color Yellow
Boiling Point 216°C
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Benzodioxane
RTECSNumber DF4728000
TSCA No