missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzodioxane-5-carboxylic Acid 98.0+%, TCI America™

Catalog No. B38645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38645G 5 g
B386425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38645G Supplier TCI America Supplier No. B38645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4442-53-9
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00239415
InChI Key VCLSWKVAHAJSFL-UHFFFAOYSA-N
Synonym 1,4-benzodioxan-5-carboxylic acid, 2,3-dihydrobenzo b 1,4 dioxine-5-carboxylic acid, 2,3-dihydro-1,4-benzodioxin-5-carboxylic acid, 1,4-benzodioxane-5-carboxylic acid, 2,3-dihydro-benzo 1,4 dioxine-5-carboxylic acid, 2,3-ethylenedioxybenzoic acid, 2,3-dihydro-1,4-benzodioxane-5-carboxylic acid, 2,3-dihydrobenzo 1,4 dioxine-5-carboxylic acid, 1,4-benzodioxin-5-carboxylic acid, 2,3-dihydro, 2h,3h-benzo e 1,4-dioxin-5-carboxylic acid
PubChem CID 78184
IUPAC Name 2,3-dihydro-1,4-benzodioxine-5-carboxylic acid
SMILES C1COC2=C(C=CC=C2O1)C(=O)O

Specifications

Melting Point 192°C
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzodioxane-5-carboxylic Acid