missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediboronic Acid Bis(pinacol) Ester 98.0+%, TCI America™

Catalog No. B463325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B463325G 25 g
B46335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B463325G Supplier TCI America Supplier No. B463325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99770-93-1
Molecular Formula C18H28B2O4
Molecular Weight (g/mol) 330.038
MDL Number MFCD08276852
InChI Key UOJCDDLTVQJPGH-UHFFFAOYSA-N
Synonym 1,4-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene, 1,4-benzenediboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-phenylenediboronic acid, pinacol ester, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2,2'-1,4-phenylene bis 4,4,5,5-tetramethyl, 4,4,5,5-tetramethyl-2-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1,3,2-dioxaborolane, 1,4-benzenediboronic acid dipinacol ester, amtb185, 1,4-phenylenediboronic acid pinacol ester
PubChem CID 10592495
IUPAC Name 4,4,5,5-tetramethyl-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)B3OC(C(O3)(C)C)(C)C

Specifications

Melting Point 243°C
Color White-Yellow
Quantity 25 g
Formula Weight 330.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Benzenediboronic Acid Bis(pinacol) Ester