missing translation for 'onlineSavingsMsg'
Learn More

1,4-Benzenediacrylic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAL0114106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0114106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0114106 Supplier Thermo Scientific Chemicals Supplier No. L0114106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16323-43-6 | Molecular Formula: C12H10O4 | Molecular Weight: 218.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16323-43-6
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.208
MDL Number MFCD00002698
InChI Key AAFXQFIGKBLKMC-KQQUZDAGSA-N
Synonym 1,4-benzenediacrylic acid, 1,4-phenylenediacrylic acid, p-phenylenediacrylic acid, p-benzenediacrylic acid, unii-0f7e4o8q8y, 3,3'-1,4-phenylene bis-2-propenoic acid, 3,3'-p-phenylene diacrylic acid, 2e-3-4-1e-2-carboxyeth-1-en-1-yl phenyl prop-2-enoic acid, 2'e-3,3'-benzene-1,4-diylbisprop-2-enoic acid, 3,3'-benzene-1,4-diylbisprop-2-enoic acid
PubChem CID 759280
IUPAC Name (E)-3-[4-[(E)-2-carboxyethenyl]phenyl]prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)C=CC(=O)O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 2579464
Formula Weight 218.21
Percent Purity 98%
Chemical Name or Material 1,4-Benzenediacrylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 240-399-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only