missing translation for 'onlineSavingsMsg'
Learn More

1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane

Catalog No. AAH26741MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH26741MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH26741MD Supplier Thermo Scientific Chemicals Supplier No. H26741MD

CAS: 170161-27-0 | C25H48N4O6 | 500.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170161-27-0
Molecular Formula C25H48N4O6
Molecular Weight (g/mol) 500.68
MDL Number MFCD09263315
InChI Key FIPOUUYFPSMVMX-UHFFFAOYSA-N
Synonym 1,4,8-tri-boc-1,4,8,11-tetraazacyclotetradecane, tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate, acmc-1c98h, 1,4,8-tri-t-butyloxycarbonyl-1,4,8,11-tetraazacyclotetradecane, 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylic acid tri-tert-butyl ester, 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylic acid tris 1,1-dimethylethyl ester, 1,4,8-tri-tert-butyl 1,4,8,11-tetraazacyclotetradecane-1,4,8-tricarboxylate
PubChem CID 10940041
IUPAC Name tritert-butyl 1,4,8,11-tetrazacyclotetradecane-1,4,8-tricarboxylate
SMILES CC(C)(C)OC(=O)N1CCCN(CCN(CCCNCC1)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C

Specifications

Melting Point 48°C to 54°C
Quantity 250 mg
Formula Weight 500.68
Sensitivity Very Hygroscopic
Chemical Name or Material 1,4,8-Tri-Boc-1,4,8,11-tetraazacyclotetradecane
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only