missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diphenyl-1,3-propanedione 98.0+%, TCI America™

Catalog No. D0910500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0910500G 500 g
D091025G 25 g
D0910100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D0910500G Supplier TCI America Supplier No. D0910500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-46-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00003085
InChI Key NZZIMKJIVMHWJC-UHFFFAOYSA-N
Synonym dibenzoylmethane, 1,3-diphenyl-1,3-propanedione, 2-benzoylacetophenone, phenyl phenacyl ketone, 1,3-propanedione, 1,3-diphenyl, rhodiastab 83, omega-benzoylacetophenone, dibenzoyl-methane, karenzu dk2, unii-ans7me8okc
PubChem CID 8433
ChEBI CHEBI:75417
IUPAC Name 1,3-diphenylpropane-1,3-dione
SMILES C1=CC=C(C=C1)C(=O)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 78°C
Boiling Point 221°C
Quantity 500 g
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Diphenyl-1,3-propanedione
EINECSNumber (4)-1332
RTECSNumber TZ1930000
TSCA Yes