missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid, 95+%, Thermo Scientific™

Catalog No. CC22901DE
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC22901DE 5g Amber Glass Bottle
CC22901CB 250mg Amber Glass Bottle
CC22901DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC22901DE Supplier Thermo Scientific Chemicals Supplier No. CC22901DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25252-46-4
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
InChI Key QRANSYHQSVJLHX-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carboxylic acid, 1,3-dimethylthieno 2,3-c pyrazole-5-carboxylic acid, 1h-thieno 2,3-c pyrazole-5-carboxylic acid, 1,3-dimethyl, 1h-thieno 2,3-c pyrazole-5-carboxylicacid, 1,3-dimethyl, 4fzj, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carboxylicacid, unavailable-see asdi_catno 500021541, 5-carboxy-1,3-dimethyl-1h-thieno 2,3-c pyrazole, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carboxylic acid, 1,3-dimethyl-1h-thieno 2,3-c-pyrazole-5-carboxylic acid
PubChem CID 2776541
IUPAC Name 1,3-dimethylthieno[2,3-c]pyrazole-5-carboxylic acid
SMILES CC1=NN(C2=C1C=C(S2)C(=O)O)C

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 196.23
Percent Purity 95%
Chemical Name or Material 1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carboxylic acid