missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 1, 3-Dimethyl-1H-pyrazole-5-carboxylic acid |
| Fourchette de pourcentages d’essai | 97% |
| Quantité | 1g |
| Poids de formule | 140.14 |
| Pourcentage de pureté | 97% |
Sécurité et manipulation
| Hazard Category | Warning |
| Hazard Statement | GHS P Statement Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse. |