missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-pyrazole-5-carbaldehyde, ≥90%, Thermo Scientific™

Numéro de catalogue. p-2694743
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC12504CB 250mg Amber Glass Bottle
CC12504DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC12504CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC12504CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 25016-09-5
Formule moléculaire C6H8N2O
Poids moléculaire (g/mol) 124.143
Numéro MDL MFCD02681907
Clé InChI JWYFGNVBKRJGTN-UHFFFAOYSA-N
Synonyme 1,3-dimethyl-1h-pyrazole-5-carbaldehyde, 1,3-dimethylpyrazole-5-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 1,3-dimethyl, 1,3-dimethyl-5-formyl-1h-pyrazole, 2,5-dimethyl-2h-pyrazole-3-carbaldehyde, 1,3-dimethylpyrazole-5-carboxaldehyde, 1,3-dimethyl-1h-pyrazole-5-carboxaldehyde, 2,5-dimethyl-3-pyrazolecarboxaldehyde, 1,3-dimethyl-1h-pyrazol-5-carbaldehyde, 1,3-dimethyl-1 h-pyrazole-5-carbaldehyde
PubChem CID 2776362
Nom de l’IUPAC 2,5-dimethylpyrazole-3-carbaldehyde
SOURIRES CC1=NN(C(=C1)C=O)C

Spécifications

Nom ou substance chimique 1, 3-Dimethyl-1H-pyrazole-5-carbaldehyde
CAS % minimum 89.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 90%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 124.14
Pourcentage de pureté 90%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.