missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodopropane (stabilized with Copper chip) 97.0+%, TCI America™

Catalog No. D061125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D061125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D061125G Supplier TCI America Supplier No. D061125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-31-6
Molecular Formula C3H6I2
Molecular Weight (g/mol) 295.89
MDL Number MFCD00001097
InChI Key AAAXMNYUNVCMCJ-UHFFFAOYSA-N
Synonym trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
PubChem CID 12314
IUPAC Name 1,3-diiodopropane
SMILES C(CI)CI

Specifications

Boiling Point 111°C
Quantity 25 g
Formula Weight 295.89
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Diiodopropane (stabilized with Copper chip)
RTECSNumber TY1080000
TSCA Yes