missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluorobenzene, 99%

Catalog No. AAA1438730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1438730 250 g
AAA1438709 10 g
AAA1438718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1438730 Supplier Thermo Scientific Chemicals Supplier No. A1438730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 372-18-9 | C6H4F2 | 114.10 g/mol

It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

Solubility
Soluble in water at 20°C 1.1g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 372-18-9
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000327
InChI Key UEMGWPRHOOEKTA-UHFFFAOYSA-N
Synonym m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene
PubChem CID 9741
ChEBI CHEBI:38584
IUPAC Name 1,3-difluorobenzene
SMILES FC1=CC(F)=CC=C1

Specifications

Melting Point -59°C
Density 1.16
Boiling Point 82°C to 83°C
Flash Point 2°C (36°F)
Odor Characteristic
Refractive Index 1.438
Quantity 250 g
UN Number UN1993
Beilstein 1904537
Solubility Information Soluble in water at 20°C 1.1g/L.
Formula Weight 114.09
Percent Purity 99%
Chemical Name or Material 1,3-Difluorobenzene
Show More Show Less
Hazard Category H225-H332
Hazard Statement GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-746-5
RTECSNumber CZ5652000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only