missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane 97.0+%, TCI America™

Catalog No. D16085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16085G 5 g
D160825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16085G Supplier TCI America Supplier No. D16085G

Chemical Identifiers

CAS 69304-37-6
Molecular Formula C12H28Cl2OSi2
Molecular Weight (g/mol) 315.425
MDL Number MFCD00009655
InChI Key DDYAZDRFUVZBMM-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2
PubChem CID 172404
IUPAC Name chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 2987
Formula Weight 315.43
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane [Hydroxyl Protecting Agent]
EINECSNumber (2)-3942
TSCA No