missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diamino-2-hydroxypropane, 95%, Thermo Scientific™

Numéro de catalogue. p-100070
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
Emballage:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC112220050 5g Glass bottle
AC112220250 25g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC112220050 Fournisseur Thermo Scientific Chemicals Code fournisseur 112220050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 616-29-5
Formule moléculaire C3H10N2O
Poids moléculaire (g/mol) 90.12
Numéro MDL MFCD00008142
Clé InChI UYBWIEGTWASWSR-UHFFFAOYSA-N
Synonyme 1,3-diamino-2-propanol, 1,3-diamino-2-hydroxypropane, diaminoisopropanol, 2-hydroxy-1,3-diaminopropane, 2-propanol, 1,3-diamino, 2-hydroxy-1,3-propanediamine, 1,3-diamino-2-hydroxy-propane, 2-propanol,3-diamino, acmc-1b1xv, 1,3 diamino-propan-2-ol
PubChem CID 61157
Nom de l’IUPAC 1,3-diaminopropan-2-ol
SOURIRES C(C(CN)O)N

Spécifications

Nom ou substance chimique 1, 3-Diamino-2-hydroxypropane
Nom Remarque 95%
CAS % minimum 94.0
CAS % maximal 100.0
Point d’ébullition 114°C (5.0mmHg)
Point d’éclair >110°C
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 95%
Emballage Glass bottle
Formule linéaire H2NCH2CH(OH)CH2NH2
Quantité 5g
Beilstein 04, 290
Renseignements sur la solubilité Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,insoluble in diethyl ether,benzene
Poids de formule 90.12
Pourcentage de pureté 95%
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-474-2
TSCA TSCA

RUO – Research Use Only