missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedione, 97%, may cont. up to 1% NaCl

Catalog No. AAA1110914
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1110914 25 g
AAA1110918 50 g
AAA1110922 100 g
AAA111090B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1110914 Supplier Thermo Scientific Chemicals Supplier No. A1110914
Only null left

CAS: 504-02-9 | C6H8O2 | 112.128 g/mol

It is used as a nitisinone intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a nitisinone intermediate.

Solubility
Soluble in water (155 g/L) at 20°C, alcohol, chloroform, acetone, boiling benzene, ether (slightly), carbon disulfide (slightly), and petr ether (slightly).

Notes
Air & Light Sensitive. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.

Chemical Identifiers

CAS 504-02-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001585
InChI Key HJSLFCCWAKVHIW-UHFFFAOYSA-N
Synonym 1,3-cyclohexanedione, dihydroresorcinol, 1,3-cyclohexandione, 1,3 cyclohexanedione, hydroresorcinol, resorcinol, dihydro, 1,3-cyclohexanone, unii-6uk3d2bxjt, 1,3-benzenediol, dihydro, 1,3-cyclohexane dione
PubChem CID 10434
ChEBI CHEBI:17766
IUPAC Name cyclohexane-1,3-dione
SMILES C1CC(=O)CC(=O)C1

Specifications

Melting Point 101°C to 105°C
Density 1.1
Quantity 25 g
Beilstein 385888
Sensitivity Light sensitive
Merck Index 14,3178
Solubility Information Soluble in water (155g/L) at 20°C,alcohol,chloroform,acetone,boiling benzene,ether (slightly),carbon disulfide (slightly),and petr ether (slightly).
Formula Weight 112.13
Percent Purity ≥97%
Concentration or Composition (by Analyte or Components) May contain up to 1% NaCl
Chemical Name or Material 1,3-Cyclohexanedione
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 207-980-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only