missing translation for 'onlineSavingsMsg'
Learn More

1,3-Cyclohexanedicarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. C21865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21865G 5 g
C218625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21865G Supplier TCI America Supplier No. C21865G

Chemical Identifiers

CAS 3971-31-1
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00134411
InChI Key XBZSBBLNHFMTEB-UHFFFAOYSA-N
PubChem CID 107205
IUPAC Name cyclohexane-1,3-dicarboxylic acid
SMILES C1CC(CC(C1)C(=O)O)C(=O)O

Specifications

Melting Point 149°C
Color White
Quantity 5 g
Formula Weight 172.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Cyclohexanedicarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-2685
TSCA Yes