missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chloroanilino)-4-phenylphthalazine 98.0+%, TCI America™

Catalog No. C288420MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C288420MG 20 mg
C2884100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C288420MG Supplier TCI America Supplier No. C288420MG
Only null left

Chemical Identifiers

CAS 78351-75-4
Molecular Formula C20H14ClN3
Molecular Weight (g/mol) 331.803
MDL Number MFCD00209838
InChI Key CEHQLKSLMFIHBF-UHFFFAOYSA-N
Synonym MY-5445, 1-(3-Chlorophenyl)amino-4-phenylphthalazine
PubChem CID 1348
IUPAC Name N-(3-chlorophenyl)-4-phenylphthalazin-1-amine
SMILES C1=CC=C(C=C1)C2=NN=C(C3=CC=CC=C32)NC4=CC(=CC=C4)Cl

Specifications

Color Yellow
Quantity 20 mg
Formula Weight 331.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Chloroanilino)-4-phenylphthalazine