missing translation for 'onlineSavingsMsg'
Learn More

1,3-Butanediol 99.0+%, TCI America™

Catalog No. B37705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37705G 5 g
B377025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37705G Supplier TCI America Supplier No. B37705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-88-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00004554
InChI Key PUPZLCDOIYMWBV-UHFFFAOYSA-N
Synonym 1,3-butanediol, 1,3-butylene glycol, 1,3-dihydroxybutane, methyltrimethylene glycol, 1,3-butandiol, 1-methyl-1,3-propanediol, beta-butylene glycol, 1,3 butylene glycol, 1,3-butylenglykol, rs-1,3-butandiol
PubChem CID 7896
ChEBI CHEBI:52683
IUPAC Name butane-1,3-diol
SMILES CC(CCO)O

Specifications

Color Yellow
Boiling Point 207°C
Quantity 5 g
Formula Weight 90.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Butanediol [for Biochemical Research]
EINECSNumber (2)-0235
RTECSNumber EK0440000
TSCA Yes