missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(benzyloxy)-2-propanol 96.0+%, TCI America™

Catalog No. B21105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21105G 5 g
B211025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21105G Supplier TCI America Supplier No. B21105G

Chemical Identifiers

CAS 6972-79-8
Molecular Formula C17H20O3
Molecular Weight (g/mol) 272.34
MDL Number MFCD00010616
InChI Key ARLSYSVVBAMYKA-UHFFFAOYSA-N
Synonym 1,3-Di-O-benzylglycerol, 1,3-Dibenzyloxy-2-propanol
PubChem CID 247614
IUPAC Name 1,3-bis(benzyloxy)propan-2-ol
SMILES OC(COCC1=CC=CC=C1)COCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 272.34
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(benzyloxy)-2-propanol
TSCA No
Recommended Storage Refrigerator