missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(4-methoxyphenyl)thiourea 98.0+%, TCI America™

Catalog No. B44821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B44821G Supplier TCI America Supplier No. B44821G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1227-45-8
Molecular Formula C15H16N2O2S
Molecular Weight (g/mol) 288.365
MDL Number MFCD00025792
InChI Key RGRJEERVXALLTH-UHFFFAOYSA-N
PubChem CID 706970
IUPAC Name 1,3-bis(4-methoxyphenyl)thiourea
SMILES COC1=CC=C(C=C1)NC(=S)NC2=CC=C(C=C2)OC

Specifications

Melting Point 188°C
Color White
Quantity 1 g
Formula Weight 288.37
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(4-methoxyphenyl)thiourea
RTECSNumber YS2750000
TSCA No