missing translation for 'onlineSavingsMsg'
Learn More

1,3-Bis(3-aminopropyl)tetramethyldisiloxane 83.0+%, TCI America™

Catalog No. B15525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15525G 5 g
B155225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15525G Supplier TCI America Supplier No. B15525G
Only null left

Chemical Identifiers

CAS 2469-55-8
Molecular Formula C10H28N2OSi2
Molecular Weight (g/mol) 248.52
MDL Number MFCD00039786
InChI Key GPXCORHXFPYJEH-UHFFFAOYSA-N
Synonym 1,3-bis 3-aminopropyl tetramethyldisiloxane, 3,3'-1,1,3,3-tetramethyldisiloxane-1,3-diyl bis propan-1-amine, 1,3-bis 3-aminopropyl-1,1,3,3-tetramethyldisiloxane, tetramethyl-1,3-bis 3-aminopropyl disiloxane, 1-propanamine, 3,3'-1,1,3,3-tetramethyl-1,3-disiloxanediyl bis, oxybis 3-aminopropyl dimethylsilane, bis 3-aminopropyl tetramethyl disiloxane, silane, oxybis 3-aminopropyl dimethyl, bis gamma-aminopropyl tetramethyldisiloxane, bis 3-aminopropyl tetramethyldisiloxane
PubChem CID 17181
IUPAC Name (3-aminopropyl)({[(3-aminopropyl)dimethylsilyl]oxy})dimethylsilane
SMILES C[Si](C)(CCCN)O[Si](C)(C)CCCN

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
UN Number 2735
Formula Weight 248.52
Percent Purity ≥83.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Bis(3-aminopropyl)tetramethyldisiloxane [Monomer for silicon modified polyamides]
EINECSNumber (7)-0453
RTECSNumber JM9235000
TSCA Yes