missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzenedisulfonyl Chloride 98.0+%, TCI America™

Catalog No. B08485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08485G 5 g
B084825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08485G Supplier TCI America Supplier No. B08485G

Chemical Identifiers

CAS 585-47-7
Molecular Formula C6H4Cl2O4S2
Molecular Weight (g/mol) 275.11
MDL Number MFCD00041879
InChI Key ALIQZUMMPOYCIS-UHFFFAOYSA-N
Synonym 1,3-benzenedisulfonyl chloride, benzene-1,3-disulfonyl dichloride, 1,3-benzenedisulfonyl dichloride, m-benzenedisulfonyl chloride, m-benzenedisulfonyl dichloride, m-phenylenedisulfonyl chloride, 1,3-benzenedisulfonyldichloride, 3-chlorosulfonylbenzenesulfonyl chloride, benzene-1,3-di sulphonyl chloride, acmc-209m5a
PubChem CID 96240
IUPAC Name benzene-1,3-disulfonyl dichloride
SMILES ClS(=O)(=O)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

Melting Point 62°C
Boiling Point 145°C
Quantity 5 g
UN Number 3261
Formula Weight 275.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Benzenedisulfonyl Chloride
EINECSNumber (3)-1899
TSCA Yes
Recommended Storage Refrigerator