missing translation for 'onlineSavingsMsg'
Learn More

1,3-Adamantanedicarboxylic Acid 97.0+%, TCI America™

Catalog No. A13585G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13585G 5 g
A135825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13585G Supplier TCI America Supplier No. A13585G

Chemical Identifiers

CAS 39269-10-8
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00167790
InChI Key PAVQGHWQOQZQEH-UHFFFAOYSA-L
Synonym 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid
PubChem CID 64339
IUPAC Name adamantane-1,3-dicarboxylate
SMILES [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 224.26
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Adamantanedicarboxylic Acid
RTECSNumber YD1994000
TSCA No