missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trithiane 98.0+%, TCI America™

Catalog No. T053225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T053225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T053225G Supplier TCI America Supplier No. T053225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 291-21-4
Molecular Formula C3H6S3
Molecular Weight (g/mol) 138.261
MDL Number MFCD00006653
InChI Key LORRLQMLLQLPSJ-UHFFFAOYSA-N
Synonym s-trithiane, trithioformaldehyde, sym-trithian, thioform, trimethylene trisulfide, 1,3,5-trithiacyclohexane, trimethylentrisulfid, sym-trithiane, formaldehyde, thio-, trimer, thioform czech
PubChem CID 9264
ChEBI CHEBI:39196
IUPAC Name 1,3,5-trithiane
SMILES C1SCSCS1

Specifications

Melting Point 216°C
Color White-Yellow
Quantity 25 g
Formula Weight 138.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Trithiane
RTECSNumber YL8225000
TSCA Yes