Learn More
1,3,5-Tris(trifluoromethyl)benzene 98.0+%, TCI America™
Supplier:  TCI America T28305G
 View more versions of this product
Catalog No. T28305G
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
1,3,5-Tris(trifluoromethyl)benzene
Colorless
C9H3F9
5 g
ZMAUHKSOLPYPDB-UHFFFAOYSA-N
1,3,5-tris(trifluoromethyl)benzene
123085
≥98.0% (GC)
729-81-7
120°C
MFCD00054722
1993
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F
282.109
282.11
Liquid
Chemical Identifiers
729-81-7
282.109
ZMAUHKSOLPYPDB-UHFFFAOYSA-N
1,3,5-tris(trifluoromethyl)benzene
C9H3F9
MFCD00054722
123085
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(F)(F)F
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No