missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene 98.0+%, TCI America™

Supplier:  TCI America T164125G

 View more versions of this product

Catalog No. T164125G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene
186°C
C12H15Br3
25 g
2,4,6-Tris(bromomethyl)mesitylene
CC1=C(C(=C(C(=C1CBr)C)CBr)C)CBr
398.964
398.96
Crystalline Powder
21988-87-4
White-Yellow
MFCD00192554
3261
BHIFXIATEXVOQA-UHFFFAOYSA-N
1,3,5-tris(bromomethyl)-2,4,6-trimethylbenzene
89140
≥98.0% (T)

Chemical Identifiers

21988-87-4
398.964
BHIFXIATEXVOQA-UHFFFAOYSA-N
89140
CC1=C(C(=C(C(=C1CBr)C)CBr)C)CBr
C12H15Br3
MFCD00192554
2,4,6-Tris(bromomethyl)mesitylene
1,3,5-tris(bromomethyl)-2,4,6-trimethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions