missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene 98.0+%, TCI America™

Catalog No. p-7136886
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16415G 5 g
T164125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16415G Supplier TCI America Supplier No. T16415G
Only null left

Chemical Identifiers

CAS 21988-87-4
Molecular Formula C12H15Br3
Molecular Weight (g/mol) 398.964
MDL Number MFCD00192554
InChI Key BHIFXIATEXVOQA-UHFFFAOYSA-N
Synonym 2,4,6-Tris(bromomethyl)mesitylene
PubChem CID 89140
IUPAC Name 1,3,5-tris(bromomethyl)-2,4,6-trimethylbenzene
SMILES CC1=C(C(=C(C(=C1CBr)C)CBr)C)CBr

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 398.96
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(bromomethyl)-2,4,6-trimethylbenzene