accessibility menu, dialog, popup

UserName

CH3BrCH3BrH3CBr

CAS RN 21988-87-4

IUPAC Name: 1,3,5-tris(bromomethyl)-2,4,6-trimethylbenzene
Synonyms: 1,3,5-tris(bromomethyl)-2,4,6-trimethylbenzene
Molecular Weight (g/mol): 398.96
Molecular Formula: C12H15Br3
InChi Key: BHIFXIATEXVOQA-UHFFFAOYSA-N
SMILES: CC1=C(CBr)C(C)=C(CBr)C(C)=C1CBr
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results