missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tris(4'-fluorobiphenyl-4-yl)benzene 95.0+%, TCI America™

Catalog No. T3462200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3462200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3462200MG Supplier TCI America Supplier No. T3462200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 372956-40-6
Molecular Formula C42H27F3
Molecular Weight (g/mol) 588.673
InChI Key KMIGKTQTBLIAJV-UHFFFAOYSA-N
Synonym 4,4′′′′-Difluoro-5′′-(4′-fluoro-[1,1′-biphenyl]-4-yl)-1,1′:4′,1′′:3′′,1′′′:4′′′,1′′′′-quinquephenyl
PubChem CID 11467407
IUPAC Name 1,3,5-tris[4-(4-fluorophenyl)phenyl]benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)F)C3=CC(=CC(=C3)C4=CC=C(C=C4)C5=CC=C(C=C5)F)C6=CC=C(C=C6)C7=CC=C(C=C7)F

Specifications

Melting Point 230°C
Color White-Yellow
Quantity 200 mg
Formula Weight 588.67
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(4′-fluorobiphenyl-4-yl)benzene