missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethylhexahydro-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7136767
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T142225ML 25 mL
T1422500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T142225ML Supplier TCI America Supplier No. T142225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-74-7
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.21
MDL Number MFCD00006057
InChI Key DPMZXMBOYHBELT-UHFFFAOYSA-N
Synonym Hexahydro-1,3,5-trimethyl-1,3,5-triazine
PubChem CID 7952
IUPAC Name 1,3,5-trimethyl-1,3,5-triazinane
SMILES CN1CN(C)CN(C)C1

Specifications

Color Yellow
Boiling Point 65°C
Quantity 25 mL
UN Number 1993
Formula Weight 129.21
Percent Purity ≥98.0% (GC,T)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,3,5-Trimethylhexahydro-1,3,5-triazine
RTECSNumber XY9300000
TSCA Yes