missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole 98.0+%, TCI America™

Catalog No. T3409200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3409200MG 200 mg
T34091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3409200MG Supplier TCI America Supplier No. T3409200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 844891-04-9
Molecular Formula C12H21BN2O2
Molecular Weight (g/mol) 236.12
MDL Number MFCD06659062
InChI Key IZNGYNMIIVJWSO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3,5-trimethyl-1h-pyrazole-4-boronic acid pinacol ester, 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3,5-trimethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1,3,5-trimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3,5-trimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, pubchem19369, 1,3,5-trimethyl-1h-pyrazole-4-boronic pinacol ester, 1,3,5-trimethylpyrazole-4-boronic acid pinacol ester
PubChem CID 2795311
IUPAC Name 1,3,5-trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES CN1N=C(C)C(B2OC(C)(C)C(C)(C)O2)=C1C

Specifications

Melting Point 101°C
Color White
Quantity 200 mg
Formula Weight 236.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole