missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester, 95%

Catalog No. AAH32930MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32930MD 250 mg
AAH3293003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32930MD Supplier Thermo Scientific Chemicals Supplier No. H32930MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 844891-04-9 | C12H21BN2O2 | 236.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 844891-04-9
Molecular Formula C12H21BN2O2
Molecular Weight (g/mol) 236.12
MDL Number MFCD06659062
InChI Key IZNGYNMIIVJWSO-UHFFFAOYSA-N
Synonym 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3,5-trimethyl-1h-pyrazole-4-boronic acid pinacol ester, 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3,5-trimethyl-1h-pyrazole-4-boronic acid, pinacol ester, 1,3,5-trimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3,5-trimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3,5-trimethyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-pyrazole, pubchem19369, 1,3,5-trimethyl-1h-pyrazole-4-boronic pinacol ester, 1,3,5-trimethylpyrazole-4-boronic acid pinacol ester
PubChem CID 2795311
IUPAC Name 1,3,5-trimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CN1N=C(C)C(B2OC(C)(C)C(C)(C)O2)=C1C

Specifications

Quantity 250 mg
Formula Weight 236.12
Percent Purity 95%
Chemical Name or Material 1,3,5-Trimethyl-1H-pyrazole-4-boronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only