missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Triethylbenzene, 95%

Catalog No. AAH6087514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6087514 25 g
AAH6087506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6087514 Supplier Thermo Scientific Chemicals Supplier No. H6087514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 102-25-0 | C12H18 | 162.28 g/mol

,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
,3,5-Triethylbenzene was used as a supramoelcular template to organize molecular-recognition elements. It was also used to synthesize a series of di- and trinucleating ligands.

Solubility
Insoluble in water. Soluble in alcohol, ether.

Notes
Incompatible with strong oxidizing agents. Store under room temperature.

Chemical Identifiers

CAS 102-25-0
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00009261
InChI Key WJYMPXJVHNDZHD-UHFFFAOYSA-N
Synonym benzene, 1,3,5-triethyl, unii-02k328wwzq, 1,5-triethylbenzene, pubchem19794, benzene,3,5-triethyl, 1,3,5-triethyl benzene, ksc493q9l, wjympxjvhndzhd-uhfffaoysa, 1,3,5-triethylbenzene, 1,3,5-triethylbenzene, technical grade
PubChem CID 7602
IUPAC Name 1,3,5-triethylbenzene
SMILES CCC1=CC(CC)=CC(CC)=C1

Specifications

Density 0.862
Boiling Point 215°C
Flash Point 76°C (169°F)
Refractive Index 1.495
Quantity 25 g
UN Number UN3082
Beilstein 2039481
Solubility Information Insoluble in water. Soluble in alcohol,ether.
Formula Weight 162.27
Percent Purity 95%
Chemical Name or Material 1,3,5-Triethylbenzene
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 203-017-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only