missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tribromobenzene 98.0+%, TCI America™

Catalog No. T03475G
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03475G 5 g
T034725G 25 g
T0347250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T03475G Supplier TCI America Supplier No. T03475G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-39-1
Molecular Formula C6H3Br3
Molecular Weight (g/mol) 314.80
MDL Number MFCD00000080
InChI Key YWDUZLFWHVQCHY-UHFFFAOYSA-N
Synonym benzene, 1,3,5-tribromo, 1,3,5-tribromo-benzene, unii-o3td0u1oaq, o3td0u1oaq, paragos 530416, 1,5-tribromobenzene, pubchem9590, 1,3,5-tribromobezene, benzene,3,5-tribromo, acmc-20a11o
PubChem CID 12279
IUPAC Name 1,3,5-tribromobenzene
SMILES BrC1=CC(Br)=CC(Br)=C1

Specifications

Melting Point 122°C
Boiling Point 271°C
Quantity 5 g
Formula Weight 314.80
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tribromobenzene
RTECSNumber DC1957200
TSCA Yes