missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Tri(9H-carbazol-9-yl)benzene (purified by sublimation) 98.0+%, TCI America™

Catalog No. p-7137028
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T19341G Supplier TCI America Supplier No. T19341G

Chemical Identifiers

CAS 148044-07-9
Molecular Formula C42H27N3
Molecular Weight (g/mol) 573.70
MDL Number MFCD03844810
InChI Key DVNOWTJCOPZGQA-UHFFFAOYSA-N
PubChem CID 22218951
IUPAC Name 9-[3,5-bis(9H-carbazol-9-yl)phenyl]-9H-carbazole
SMILES C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC(=CC(=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 330°C
Color White
Quantity 1 g
Formula Weight 573.70
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tri(9H-carbazol-9-yl)benzene (purified by sublimation)