missing translation for 'onlineSavingsMsg'
Learn More

1-(3,5-Difluoro-2-hydroxyphenyl)ethan-1-one, Thermo Scientific™

Catalog No. BTB04131EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB04131EA 10 g Amber Glass Bottle
BTB04131DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB04131EA Supplier Thermo Scientific Chemicals Supplier No. BTB04131EA

Chemical Identifiers

CAS 140675-42-9
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.131
MDL Number MFCD00042476
InChI Key MCDJUVXLLXTCFP-UHFFFAOYSA-N
Synonym 1-3,5-difluoro-2-hydroxyphenyl ethanone, 1-3,5-difluoro-2-hydroxyphenyl ethan-1-one, 3',5'-difluoro-2'-hydroxyacetophenone, 3,5-difluoro-2-hydroxyacetophenone, 2-acetyl-4,6-difluorophenol, ethanone,1-3,5-difluoro-2-hydroxyphenyl, 1-acetyl-3,5-difluoro-2-hydroxybenzene, acmc-1byqa, 1-3,5-difluoro-2-hydroxyphenyl-ethanone, 1-3,5-difluoro-2-hydroxy-phenyl-ethanone
PubChem CID 2736976
IUPAC Name 1-(3,5-difluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC(=C1O)F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 1-(3, 5-Difluoro-2-hydroxyphenyl)ethan-1-one
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.