missing translation for 'onlineSavingsMsg'
Learn More

1,3,2-Dioxathiane 2-Oxide 98.0+%, TCI America™

Catalog No. D442625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D442625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D442625G Supplier TCI America Supplier No. D442625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4176-55-0
Molecular Formula C3H6O3S
Molecular Weight (g/mol) 122.138
MDL Number MFCD01682880
InChI Key LOURZMYQPMDBSR-UHFFFAOYSA-N
Synonym 1,3-Propylene Sulfite, Trimethylene Sulfite, 1,3-Propanediol Cyclic Sulfite
PubChem CID 77839
IUPAC Name 1,3,2-dioxathiane 2-oxide
SMILES C1COS(=O)OC1

Specifications

Color Colorless
Boiling Point 54°C
Quantity 25 g
Formula Weight 122.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3,2-Dioxathiane 2-Oxide