missing translation for 'onlineSavingsMsg'
Learn More

1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole 98.0+%, TCI America™

Catalog No. T25901G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25901G 1 g
T25905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25901G Supplier TCI America Supplier No. T25901G

Chemical Identifiers

CAS 113306-55-1
Molecular Formula C12H17N3O3Si
Molecular Weight (g/mol) 279.37
MDL Number MFCD11846123
InChI Key BJUJSLXVBATBSB-UHFFFAOYSA-N
Synonym Teoc-OBt
PubChem CID 13892319
IUPAC Name 1H-1,2,3-benzotriazol-1-yl 2-(trimethylsilyl)ethyl carbonate
SMILES C[Si](C)(C)CCOC(=O)ON1N=NC2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 279.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-[2-(Trimethylsilyl)ethoxycarbonyloxy]benzotriazole