missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog No. AAH2703603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2703603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2703603 Supplier Thermo Scientific Chemicals Supplier No. H2703603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 903550-26-5 | Molecular Formula: C14H23BN2O3 | Molecular Weight: 278.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 903550-26-5
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD09037501
InChI Key ZZRFDLHBMBHJTI-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-2-tetrahydropyranyl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-y-1h-pyrazole, 1-tetrahydropyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole
PubChem CID 11587208
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2C3CCCCO3

Specifications

Quantity 1 g
Formula Weight 278.16
Percent Purity 95%
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only